EMAIL THIS PAGE TO A FRIEND
American journal of perinatology
Association of Antegrade Pulmonary Artery Diastolic Velocity with Spontaneous Closure of the Patent Ductus Arteriosus in Extremely Low-Birth-Weight Infants.
PMID 26058372
Abstract
This study aims to determine echocardiographic parameters associated with spontaneous patent ductus arteriosus (PDA) closure in extremely low-birth-weight (ELBW) infants. Retrospective demographic review and analysis of echocardiograms from 189 ELBW infants with suspected and confirmed hemodynamically significant PDA identified on an initial echocardiogram was performed. Comparison of echocardiographic parameters was made between infants with spontaneous closure versus those who received treatment. The mean birth weight (787 ± 142 vs. 724 ± 141 g, p = 0.04) and gestational age (27.4 ± 2.8 vs. 26.2 ± 1.6 weeks, p = 0.03) were higher in the spontaneous closure versus the treatment group. Antegrade pulmonary artery (PA) diastolic velocity was lower in infants with spontaneous PDA closure versus those who received treatment (0.15 ± 0.06 vs. 0.22 ± 0.12 m/s, p = 0.009). Heavier and more mature ELBW infants with a lower antegrade PA diastolic velocity were likely to have spontaneous closure of the PDA.