EMAIL THIS PAGE TO A FRIEND
Analytical chemistry
Cleavable Molecular Beacon for Hg(2+) Detection Based on Phosphorothioate RNA Modifications.
PMID 26060876
Abstract
Mercury is a highly toxic heavy metal, and detection of Hg(2+) by biosensors has attracted extensive research interest in the past decade. In particular, a number of DNA-based sensing strategies have been developed. Well-known examples include thymine-Hg(2+) interactions and Hg(2+)-activated DNAzymes. However, these mechanisms are highly dependent on buffer conditions or require hybridization with another DNA strand. Herein, we report a new mechanism based on Hg(2+)-induced cleavage of phosphorothioate (PS) modified RNA. Among the various metal ions tested, Hg(2+) induced the most significant cleavage (∼16%), while other metals cleaved less than 2% of the same substrate. The uncleaved substrate undergoes desulfurization in the presence of Hg(2+). This cleavage reaction yields a similar amount of product from pH 3.5 to 7 and in the temperature range between 20 and 90 °C. Various PS RNA junctions can be cleaved with a similar efficiency, but PS DNA junctions cannot be cleaved. A molecular beacon containing three PS RNA modifications is designed, detecting Hg(2+) down to 1.7 nM with excellent selectivity. This sensor can also detect Hg(2+) in the Lake Ontario water sample, although its response is significantly masked by fish tissues.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
229393
Aluminum chloride hydrate, 99.999% trace metals basis
AlCl3 · xH2O
449644
Barium chloride, anhydrous, beads, −10 mesh, 99.999% trace metals basis
BaCl2
449652
Barium chloride, anhydrous, beads, −10 mesh, 99.95% trace metals basis
BaCl2
202738
Barium chloride, 99.999% trace metals basis
BaCl2
342920
Barium chloride, 99.9% trace metals basis
BaCl2
529575
Cadmium chloride hydrate, 99.995% trace metals basis
CdCl2 · xH2O
208299
Cadmium chloride hydrate, 98%
CdCl2 · xH2O
C5080
Calcium chloride dihydrate, BioXtra, ≥99.0%
CaCl2 · 2H2O
C7902
Calcium chloride dihydrate, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
CaCl2 · 2H2O
C3306
Calcium chloride dihydrate, for molecular biology, ≥99.0%
CaCl2 · 2H2O
21097
Calcium chloride dihydrate, BioUltra, for molecular biology, ≥99.5% (KT)
CaCl2 · 2H2O
202983
Cerium(III) chloride heptahydrate, 99.999% trace metals basis
CeCl3 · 7H2O
228931
Cerium(III) chloride heptahydrate, 99.9% trace metals basis
CeCl3 · 7H2O
22300
Cerium(III) chloride heptahydrate, purum p.a., ≥98.0% (AT)
CeCl3 · 7H2O
203025
Cesium chloride, ≥99.999% trace metals basis
ClCs
449733
Cesium chloride, anhydrous, beads, −10 mesh, 99.999% trace metals basis
ClCs
C3139
Cesium chloride, optical grade, ≥99.5% trace metals basis
ClCs
C3011
Cesium chloride, Grade I, ≥99.0%
ClCs
562599
Cesium chloride, 99.99% trace metals basis
ClCs
C3032
Cesium chloride, for molecular biology, ≥99% (silver nitrate titration)
ClCs
C4036
Cesium chloride, for molecular biology, ≥98%
ClCs
C3309
Cesium chloride, BioXtra, ≥99.5% (titration)
ClCs
C6914
Cesium chloride, Grade II, ≥98%
ClCs
20966
Cesium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
ClCs
C8661
Cobalt(II) chloride hexahydrate, BioReagent, suitable for cell culture, suitable for insect cell culture
Cl2Co · 6H2O
C2911
Cobalt(II) chloride hexahydrate, plant cell culture tested
Cl2Co · 6H2O
C3279
Copper(II) chloride dihydrate, powder, BioReagent, suitable for cell culture
Cl2Cu · 2H2O
459097
Copper(II) chloride dihydrate, 99.999%
Cl2Cu · 2H2O
467847
Copper(II) chloride dihydrate, ≥99.95% trace metals basis
Cl2Cu · 2H2O
427128
Gallium(III) chloride, beads, anhydrous, ≥99.999% trace metals basis
Cl3Ga