EMAIL THIS PAGE TO A FRIEND
Molecules (Basel, Switzerland)
Analysis of Flavonoids in Lotus (Nelumbo nucifera) Leaves and Their Antioxidant Activity Using Macroporous Resin Chromatography Coupled with LC-MS/MS and Antioxidant Biochemical Assays.
PMID 26060918
Abstract
Lotus (Nelumbo nucifera) leaves, a traditional Chinese medicinal herb, are rich in flavonoids. In an effort to thoroughly analyze their flavonoid components, macroporous resin chromatography coupled with HPLC-MS/MS was employed to simultaneously enrich and identify flavonoids from lotus leaves. Flavonoids extracted from lotus leaves were selectively enriched in the macroporous resin column, eluted subsequently as fraction II, and successively subjected to analysis with the HPLC-MS/MS and bioactivity assays. Altogether, fourteen flavonoids were identified, four of which were identified from lotus leaves for the first time, including quercetin 3-O-rhamnopyranosyl-(1→2)-glucopyranoside, quercetin 3-O-arabinoside, diosmetin 7-O-hexose, and isorhamnetin 3-O-arabino- pyranosyl-(1→2)-glucopyranoside. Further bioactivity assays revealed that these flavonoids from lotus leaves possess strong antioxidant activity, and demonstrate very good potential to be explored as food supplements or even pharmaceutical products to improve human health.