EMAIL THIS PAGE TO A FRIEND
Anais da Academia Brasileira de Ciencias
Competition between rice (Oryza sativa L.) and (barnyardgrass (Echinochloa crus-galli (L.) P. Beauv.) as affected by methanol foliar application.
PMID 26062115
Abstract
Pot experiment was conducted in Iran, to evaluate the effect of methanol on competition between rice (Oryza sativa) and barnyardgrass (Echinochloa crus-galli). The experiment was conducted as a randomized complete block design with a factorial treatment arrangement and three replicates. Factors were two aqueous methanol foliar applications (0, and 14% v/v) and five rice: barnyardgrass ratios (100:0, 75:25, 50:50, 25:6, and 0:100). Replacement series diagrams for aboveground dry weight illustrated that 'Shiroudi' was more competitive than barnyardgrass as averaged across methanol foliar applications. When methanol was not sprayed, the lines for 'Shiroudi' and barnyardgrass intersected at 75:25 rice: barnyardgrass ratio, but when methanol was sprayed at 14% v/v, the lines for 'Shiroudi' and barnyardgrass intersect at the left of the 75:25 rice: barnyardgrass mixture proportion. These indicate that methanol application reduced competitive ability of 'Shiroudi' against barnyardgrass for aboveground biomass accumulation. At the same time, Methanol foliar application significantly reduced the relative crowding coefficient of 'Shiroudi' while simultaneously it significantly increased the relative crowding coefficient of barnyard grass. This indicates that methanol foliar application reduced the competitive ability of 'Shiroudi' against barnyardgrass for shoot biomass accumulation. This experiment illustrated that foliar spray of aqueous methanol can not be recommended for rice under weedy conditions.