EMAIL THIS PAGE TO A FRIEND
Journal of immunology (Baltimore, Md. : 1950)
Evaluating the Role of HLA-DM in MHC Class II-Peptide Association Reactions.
PMID 26062997
Abstract
Ag presentation by MHC class II (MHC II) molecules to CD4(+) T cells plays a key role in the regulation of the adaptive immune response. Loading of antigenic peptides onto MHC II is catalyzed by HLA-DM (DM), a nonclassical MHC II molecule. The mechanism of DM-facilitated peptide loading is an outstanding problem in the field of Ag presentation. In this study, we systemically explored possible kinetic mechanisms for DM-catalyzed peptide association by measuring real-time peptide association kinetics using fluorescence polarization assays and comparing the experimental data with numerically modeled peptide association reactions. We found that DM does not facilitate peptide association by stabilizing peptide-free MHC II against aggregation. Moreover, DM does not promote transition of an inactive peptide-averse conformation of MHC II to an active peptide-receptive conformation. Instead, DM forms an intermediate with MHC II that binds peptide with faster kinetics than MHC II in the absence of DM. In the absence of peptides, interaction of MHC II with DM leads to inactivation and formation of a peptide-averse form. This study provides novel insights into how DM efficiently catalyzes peptide loading during Ag presentation.