EMAIL THIS PAGE TO A FRIEND
Disease markers
Expression of Spindle and Kinetochore-Associated Protein 1 Is Associated with Poor Prognosis in Papillary Thyroid Carcinoma.
PMID 26063960
Abstract
Spindle and kinetochore-associated protein 1 (SKA1) is one subtype of SKA, whose protein can make spindle microtubules attach steadily to the kinetochore in the middle of mitosis. At present, there are fewer researches on the relationship between SKA1 expression and tumor development. In this study, immunohistochemical analysis was used to determine the expression of SKA1 in papillary thyroid carcinoma (PTC) and adjacent tissues. We used quantitative real-time polymerase chain reaction (qRT-PCR) and Western blot analysis to further verify the results. We found that SKA1 expression was significantly higher in PTC tissues than normal adjacent tissues (P < 0.05). There existed a significant correlation among a higher SKA1 expression, including lymphoid node (P = 0.005), clinical stage (P = 0.015), and extrathyroid invasion (P = 0.004). Survival analysis showed high SKA1 expression in PTC patients more likely to relapse after surgery. High SKA1 expression is predictive of poor prognosis of PTC, implying that SKA1 may be a promising new target for targeted therapies for PTC.