EMAIL THIS PAGE TO A FRIEND
The American journal of cardiology
Meta-Analysis of the Impact on Mortality of Noninfarct-Related Artery Coronary Chronic Total Occlusion in Patients Presenting With ST-Segment Elevation Myocardial Infarction.
PMID 26068700
Abstract
Several observational studies have compared clinical outcome in patients with a co-existing noninfarct-related artery chronic total occlusion (n-IRA CTO) versus those without, suggesting increased all-cause mortality. The goal of this study was to provide a systematic review and meta-analysis evaluating the impact of the presence of an n-IRA CTO on short- and long-term mortality after primary percutaneous coronary intervention. Studies published from January 1980 to January 2014 that compared the incidence of all-cause mortality in patients with ST-segment elevation myocardial infarction with co-existing n-IRA CTO versus those without were identified using an electronic search and reviewed using meta-analytical techniques. Seven studies (5 observational studies and 2 observational analyses of randomized controlled trials) comprising 14,117 patients and 1,554 patients (11.7%) with n-IRA CTO were included. The presence of n-IRA CTO was associated with increased incidence of all-cause mortality at a median follow-up of 25.2 months (interquartile range 24 to 60) compared with no CTO (absolute risk 23.5% vs 9.0%; odds ratio [OR] 2.90, 95% confidence interval [CI] 2.09 to 4.01; p <0.0001). This finding was consistent in the analysis of studies reporting 30-day follow-up (absolute risk 17.2% vs 4.7%; OR 3.79, 95% CI 3.13 to 4.59; p <0.0001). Co-existing n-IRA CTO was also associated with increased mortality in a subanalysis of patients with multivessel disease only (absolute risk 24.2% vs 11.3%; OR 2.23, 95% CI 1.90 to 2.63; p <0.0001). In conclusion, coronary CTO in the nonculprit artery in patients presenting with ST-segment elevation myocardial infarction is associated with increased short- and long-term all-cause mortality.