EMAIL THIS PAGE TO A FRIEND
Journal of nuclear medicine : official publication, Society of Nuclear Medicine
Prognostic Factors in Patients Treated with 223Ra: The Role of Skeletal Tumor Burden on Baseline 18F-Fluoride PET/CT in Predicting Overall Survival.
PMID 26069307
Abstract
The purpose of this study was to evaluate outcome after (223)Ra dichloride therapy ((223)Ra) and to determine whether skeletal tumor burden on whole-body (18)F-fluoride PET/CT can be used as a predictive biomarker of survival in patients treated with (223)Ra. Forty-two patients with hormone-refractory prostate cancer underwent (223)Ra and a baseline fluoride PET/CT scan. Fluoride PET/CT parameters were generated, including maximum standardized uptake value (SUVmax) of the hottest lesion (hSUVmax), average SUV of disease (Mean10), and skeletal tumor burden indices of total fluoride skeletal metastatic lesion uptake (TLF10) and total volume of fluoride avid bone metastases (FTV10). Overall survival (OS) was the primary endpoint. Secondary endpoints were progression-free survival and skeletal-related event (SRE). Skeletal tumor burden indices (TLF10 and FTV10) derived from fluoride PET/CT at baseline were highly correlated and significant independent predictors of OS (P = 0.0212; hazard ratio = 5.990; 95% confidence interval = 1.306-27.475). A TLF10 cutoff value of 8,000 discriminated survivors from nonsurvivors after (223)Ra (with TLF10 values < 8,000, the median OS was not estimated, whereas with TLF10 > 8,000, the median OS was 6.67 mo). Visual analysis, Mean10, and hSUVmax were not predictors of OS or progression-free survival. Mean10 was found to be a significant univariate predictor of the odds of having an SRE (P = 0.0445; odds ratio = 1.30; 95% confidence interval = 1.006-1.681), with a Mean10 greater than 19 increasing the risk of SRE. Skeletal tumor burden on baseline fluoride PET/CT is a predictive biomarker of OS and the risk of an SRE in patients treated with (223)Ra.