EMAIL THIS PAGE TO A FRIEND
Histochemistry and cell biology
Trophoblast expression dynamics of the tumor suppressor gene gastrokine 2.
PMID 26070363
Abstract
Gastrokines (GKNs) were originally described as stomach-specific tumor suppressor genes. Recently, we identified GKN1 in extravillous trophoblasts (EVT) of human placenta. GKN1 treatment reduced the migration of the trophoblast cell line JEG-3. GKN2 is known to inhibit the proliferation, migration and invasion of gastric cancer cells and may interact with GKN1. Recently, GKN2 was detected in the placental yolk sac of mice. We therefore aimed to further characterize placental GKN2 expression. By immunohistochemistry, healthy first-trimester placenta showed ubiquitous staining for GKN2 at its early gestational stage. At later gestational stages, a more differentiated expression pattern in EVT and villous cytotrophoblasts became evident. In healthy third-trimester placenta, only EVT retained strong GKN2 immunoreactivity. In contrast, HELLP placentas showed a tendency of increased levels of GKN2 expression with a more prominent GKN2 staining in their syncytiotrophoblast. Choriocarcinoma cell lines did not express GKN2. Besides its trophoblastic expression, we found human GKN2 in fibrotic villi, in amniotic membrane and umbilical cord. GKN2 co-localized with smooth muscle actin in villous myofibroblasts and with HLA-G and GKN1 in EVT. In the rodent placenta, GKN2 was specifically located in the spongiotrophoblast layer. Thus, the gestational age-dependent and compartment-specific expression pattern of GKN2 points to a role for placental development. The syncytial expression of GKN2 in HELLP placentas might represent a reduced state of functional differentiation of the syncytiotrophoblast. Moreover, the specific GKN2 expression in the rodent spongiotrophoblast layer (equivalent to human EVT) might suggest an important role in EVT physiology.