EMAIL THIS PAGE TO A FRIEND
Neurobiology of Disease
RNA-sequencing reveals transcriptional up-regulation of Trem2 in response to bexarotene treatment.
PMID 26071899
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
105228
1,1,1,3,3,3-Hexafluoro-2-propanol, ≥99%
C3H2F6O
SML0282
Bexarotene, ≥98% (HPLC)
C24H28O2