EMAIL THIS PAGE TO A FRIEND
European Journal of Pharmaceutics and Biopharmaceutics
CURCUMIN-EUDRAGIT? E PO SOLID DISPERSION: A SIMPLE AND POTENT METHOD TO SOLVE THE PROBLEMS OF CURCUMIN.
PMID 26073546