EMAIL THIS PAGE TO A FRIEND
International journal of endocrinology
Total and High Molecular Weight Adiponectin Levels and Prediction of Cardiovascular Risk in Diabetic Patients.
PMID 26074960
Abstract
The study aimed at assessing the potential use of lower total and HMW adiponectin levels for predicting cardiovascular risk in patients with type 2 diabetes mellitus (T2DM). Concentrations of total adiponectin or high molecular weight (HMW) adiponectin decrease in association with the development of metabolic dysfunction such as obesity, insulin resistance, or T2DM. Increased adiponectin levels are associated with a lower risk for coronary heart disease. A total of 551 individuals were assessed. The first group comprised metabolically healthy participants (143 females, and 126 males) and the second group were T2DM patients (164 females, and 118 males). Both total adiponectin and HMW adiponectin in diabetic patients were significantly lower when compared with the group of metabolically healthy individuals. There was a weak monotonic correlation between HMW adiponectin levels and triglycerides levels. Binary logistic regression analysis, gender adjusted, showed a higher cardiovascular risk in diabetic persons when both total adiponectin (OR = 1.700) and HMW adiponectin (OR = 2.785) levels were decreased. A decrease in total adiponectin levels as well as a decrease in its HMW adiponectin is associated with a higher cardiovascular risk in individuals with T2DM. This association suggests that adiponectin levels may be potentially used as an epidemiological marker for cardiovascular risk in diabetic patients.