EMAIL THIS PAGE TO A FRIEND
Antimicrobial agents and chemotherapy
The agmatine-containing poly(amidoamine) polymer AGMA1 binds cell surface heparan sulfates and prevents attachment of mucosal human papillomaviruses.
PMID 26077258
Abstract
The agmatine-containing poly(amidoamine) polymer AGMA1 was recently shown to inhibit the infectivity of several viruses, including human papillomavirus 16 (HPV-16), that exploit cell surface heparan sulfate proteoglycans (HSPGs) as attachment receptors. The aim of this work was to assess the antiviral activity of AGMA1 and its spectrum of activity against a panel of low-risk and high-risk HPVs and to elucidate its mechanism of action. AGMA1 was found to be a potent inhibitor of mucosal HPV types (i.e., types 16, 31, 45, and 6) in pseudovirus-based neutralization assays. The 50% inhibitory concentration was between 0.34 μg/ml and 0.73 μg/ml, and no evidence of cytotoxicity was observed. AGMA1 interacted with immobilized heparin and with cellular heparan sulfates, exerting its antiviral action by preventing virus attachment to the cell surface. The findings from this study indicate that AGMA1 is a leading candidate compound for further development as an active ingredient of a topical microbicide against HPV and other sexually transmitted viral infections.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
284505
1,2-Dichloroethane, anhydrous, 99.8%
C2H4Cl2
07-2101
1,2-Dichloroethane, for HPLC
C2H4Cl2
151882
Deuterium oxide, 99.9 atom % D
D2O
756822
Deuterium oxide, filtered, 99.8 atom % D
D2O
617385
Deuterium oxide, 99.8 atom % D
D2O
151890
Deuterium oxide, "100%", ≥99.96 atom % D
D2O
450510
Deuterium oxide, 99.9 atom % D, contains 0.05 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
D2O
435767
Deuterium oxide, 99 atom % D
D2O
613444
Deuterium oxide, 99.9 atom % D, glass distilled
D2O
293040
Deuterium oxide, 99.9 atom % D, contains 0.75 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
D2O
343773
Deuterium oxide, 99.9 atom % D, contains 1 % (w/w) 3-(trimethylsilyl)-1-propanesulfonic acid, sodium salt (DSS)
D2O
364312
Deuterium oxide, standard, 99.98 atom %±0.01 atom % D
D2O
613428
Deuterium oxide, 70 atom % D
D2O
191701
Deuterium oxide, "100%", 99.990 atom % D
D2O
613398
Deuterium oxide, extra, 99.994 atom % D
D2O
423459
Deuterium oxide, "100%", 99.96 atom % D
D2O
441376
Deuterium oxide, 99.9 atom % D
D2O
269794
Deuterium oxide, 99.9 atom % D
D2O
P3532
Phenol Red, powder, BioReagent, suitable for cell culture
C19H14O5S
114529
Phenol Red, ACS reagent
C19H14O5S
192341
Deuterium oxide, "100%", 99.96 atom % D, purified by glass distillation
D2O