EMAIL THIS PAGE TO A FRIEND
Nutrition journal
Intake of partially defatted Brazil nut flour reduces serum cholesterol in hypercholesterolemic patients--a randomized controlled trial.
PMID 26077768
Abstract
Thyroid hormones can lower levels of atherogenic lipoproteins, and selenium is important in thyroid hormone homeostasis. We aimed to investigate the effects of a healthy diet associated with the Brazil nut (Bertholletia excelsa) in dyslipidemic and hypertensive patients. This study was a randomized, placebo-controlled, double-blind trial. Seventy-seven dyslipidemic and hypertensive patients already receiving lipid-lowering drugs received either a dietary treatment associated with partially defatted Brazil nut flour (13 g/day providing 227,5 μg of selenium/day),or with dyed cassava flour as a placebo. All patients received a personalized dietary guideline with nutritional recommendations for dyslipidemia and hypertension and were followed for 90 days. The Brazil nut group showed reductions in total cholesterol (-20.5 ± 61.2 mg/dL, P = 0.02), non HDL-cholesterol (-19.5 ± 61.2 mg/dL, P = 0.02) and Apo A-1 (-10.2 ± 26.7 mg/dL, P = 0.03) without significant alterations in the Apo B/Apo A-1 ratio. The placebo group showed a reduction in FT3 levels (-0.1 ± 0.4, P = 0.03) and increased Lp(a) levels (5.9 ± 18.0 mg/dL, P = 0.02). There were no statistical differences in blood pressure and serum lipids between Brazil nut and placebo group. Supplementation with Brazil nuts seems to favor the maintenance of FT3 levels and contributes to lipemia reduction in hypercholesterolemic and euthyroid patients. ClinicalTrials.gov Identifier NCT01990391.