EMAIL THIS PAGE TO A FRIEND
Neurobiology of learning and memory
The retrosplenial cortex is involved in the formation of memory for context and trace fear conditioning.
PMID 26079095
Abstract
The retrosplenial cortex (RSC) is known to play a role in the retrieval of context memory, but its involvement in memory formation and consolidation is unclear. To better characterize the role of the RSC, we tested its involvement in the formation and retrieval of memory for trace fear conditioning, a task that requires the association of two cues separated by an empty period of time. We have previously shown that trace fear extinction requires the RSC (Kwapis, Jarome, Lee, Gilmartin, & Helmstetter, 2014) and have hypothesized that trace memory may be stored in a distributed cortical network that includes prelimbic and retrosplenial cortices (Kwapis, Jarome, & Helmstetter, 2015). Whether the RSC participates in acquiring and storing cued trace fear, however, is currently unknown. Here, we demonstrate that blocking protein synthesis in the RSC before, but not after acquisition impairs rats' memory for trace CS and context fear without affecting memory for the CS in standard delay fear conditioning. We also show that NMDA receptor blockade in the RSC transiently impairs memory retrieval for trace, but not delay memory. The RSC therefore appears to critically contribute to formation of trace and context fear memory in addition to its previously recognized role in context memory retrieval.