EMAIL THIS PAGE TO A FRIEND
The Journal of Physical Chemistry B
Mechanism of Formation of Copper(II) Chlorocomplexes Revealed by Transient Absorption Spectroscopy and DFT/TDDFT-Calculations.
PMID 26079181