EMAIL THIS PAGE TO A FRIEND
Environmental Science and Pollution Research International
The suitability of extraction solutions to assess bioaccessible trace metal fractions in airborne particulate matter: a comparison of common leaching agents.
PMID 26081774