EMAIL THIS PAGE TO A FRIEND
Molecules (Basel, Switzerland)
Volatile Compounds from Grape Skin, Juice and Wine from Five Interspecific Hybrid Grape Cultivars Grown in Québec (Canada) for Wine Production.
PMID 26083035
Abstract
Developed from crosses between Vitis vinifera and North American Vitis species, interspecific hybrid grape varieties are becoming economically significant in northern areas, where they are now extensively grown for wine production. However, the varietal differences between interspecific hybrids are not well defined, nor are the relationships between hybrid grape and wine composition, which causes significant drawbacks in the development of viticulture and winemaking of northern wines. In an effort to increase our understanding of interspecific hybrids, we have characterized the free volatile compounds profiles of berries (juice and skin) and wines of five red hybrid varieties (Frontenac, Marquette, Maréchal Foch, Sabrevois and St. Croix) grown in Québec (Canada), using GC-MS(TOF)-SPME. In grapes and wines, significantly higher levels of C6 and other fatty acid degradation products (FADP) were found in Frontenac, Maréchal Foch and Marquette. Terpenes were primarily located in the skin, with Marquette showing the highest level for these compounds. Both the level of terpenes and the level of FADP in grape were strongly correlated with their respective levels in wine, as demonstrated by the redundancy analyses. Nonanal, (E,Z)-2,6-nonadienal, β-damascenone, ethyl octanoate and isoamyl acetate showed the highest OAVs in the wines of the studied varieties.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E34102
(−)-Ethyl L-lactate, 98%
C5H10O3
62139
(−)-Linalool, ≥95.0% (sum of enantiomers, GC)
C10H18O
183164
(R)-(+)-Limonene, 97%
C10H16
W263303
(R)-(+)-Limonene, ≥93%
C10H16
H2805
1-Heptanol, 98%
C7H16O
51778
1-Heptanol, ≥99.5% (GC)
C7H16O
471402
1-Hexanol, anhydrous, ≥99%
C6H14O
52828
1-Hexanol, ReagentPlus®, ≥99.5% (GC)
C6H14O
W280518
1-Octen-3-ol, ≥98%, FCC, FG
C8H16O
O5284
1-Octen-3-ol, 98%
C8H16O
W280523
1-Octen-3-ol, natural, ≥95%, FG
C8H16O
777064
1-Octen-3-one, 96%
C8H14O
294829
2-Methyl-1-propanol, anhydrous, 99.5%
C4H10O
58448
2-Methyl-1-propanol, BioUltra, for molecular biology, ≥99.5% (GC)
C4H10O
O4709
2-Octanone, reagent grade, 98%
C8H16O
W280208
2-Octanone, ≥98%, FG
C8H16O
W280220
2-Octanone, natural, 98%, FG
C8H16O
77861
2-Phenylethanol, ≥99.0% (GC)
C8H10O
W309303
2-Undecanone, ≥98%, FCC, FG
C11H22O
U1303
2-Undecanone, 99%
C11H22O
W309311
2-Undecanone, natural, FCC, FG
C11H22O
W329118
4-Hydroxybutanoic acid lactone, ≥98%, FCC, FG
C4H6O2
W200808
Acetoin, primarily dimer, ≥95%, FG
C4H8O2
W200832
Acetoin, natural, ≥95%, FG
C4H8O2
A17951
Acetoin, May exist as crystalline dimer
C4H8O2
362999
Benzyl-2,3,4,5,6-d5 alcohol, 98 atom % D
C7D5H3O
268909
cis-3,7-Dimethyl-2,6-octadien-1-ol, 97%
C10H18O
W277002
cis-3,7-Dimethyl-2,6-octadien-1-ol, ≥97%, FCC, FG
C10H18O
G1951
D-Gluconic acid solution, 49-53 wt. % in H2O
C6H12O7
D7384
Decanal, ≥98% (GC), liquid
C10H20O