EMAIL THIS PAGE TO A FRIEND
Nucleic acids symposium series
A phosphorbisamidite approach to the synthesis of oligodeoxyribonucleotides and their analogues.
PMID 2608468
Abstract
Utilities of deoxyribonucleoside 3'-O-phosphorbisdiethylamidites in the synthesis of oligodeoxyribonucleotides and their analogues are described.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
362565
Bis(diethylamino)chlorophosphine, 97%
C8H20ClN2P
253189
Tris(diethylamino)phosphine, 97%
C12H30N3P