EMAIL THIS PAGE TO A FRIEND
Nucleic acids symposium series
The introduction of reporter groups at multiple and/or specific sites in DNA containing phosphorothioate diesters.
PMID 2608472
Abstract
DNA fragments containing phosphorothioate diesters provide nucleophilic sites which are amenable to labeling by spin labels or fluorophores. Selecting the position for an individual phosphorothioate diester allows highly specific placement of the reporter group. The substitution of a phosphorothioate diester for each and every internucleotidic phosphodiester allows the incorporation of multiple reporter groups; ideally one for each nucleoside residue. With the use of multiple fluorophores a post-assay fluorescent labeling technique has been developed which allows the detection of DNA fragments with the "naked-eye" in the low femtomolar (10(-15) moles) range.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
253421
3-(2-Iodoacetamido)-PROXYL, free radical
C10H18IN2O2