EMAIL THIS PAGE TO A FRIEND
Acta chimica Slovenica
An axial chirality and disorder of positional isomers in a crystal of highly cytotoxic 3-acetyl-1,3-bis(2-chloro-4-nitrophenyl)-1E-triazene.
PMID 26085419
Abstract
Biologically active 4-nitro-substituted 1,3-diaryltriazene, a chemical analogue of 1,3-bis(4'-amidinophenyl)-triazene-berenil®, belongs to the novel, chemically modified class of potent antitumor agents. Its structural characterization by X-ray analysis and 1H NMR spectroscopy is performed to determine molecular overall conformation in view of its possible interaction to DNA.