EMAIL THIS PAGE TO A FRIEND
NMR in biomedicine
Monitoring dynamic alterations in calcium homeostasis by T1-mapping manganese-enhanced MRI (MEMRI) in the early stage of small intestinal ischemia-reperfusion injury.
PMID 26086648
Abstract
Manganese-enhanced MRI studies have proven to be useful in monitoring physiological activities associated with calcium ions (Ca(2+)) due to the paramagnetic property of the manganese ion (Mn(2+)), which makes it an excellent probe of Ca(2+) . In this study, we developed a method in which a Mn(2+)-enhanced T1 -map MRI could enable the monitoring of Ca(2+) influx during the early stages of intestinal ischemia-reperfusion (I/R) injury. The Mn(2+) infusion protocol was optimized by obtaining dose-dependent and time-course wash-out curves using a Mn(2+)-enhanced T1-map MRI of rabbit abdomens following an intravenous infusion of 50 mmol/l MnCl2 (5-10 nmol/g body weight (BW)). In the rabbit model of intestinal I/R injury, T1 values were derived from the T1 maps in the intestinal wall region and revealed a relationship between the dose of the infused MnCl2 and the intestinal wall relaxation time. Significant Mn(2+) clearance was also observed over time in control animals after the infusion of Mn(2+) at a dose of 10 nmol/g BW. This technique was also shown to be sensitive enough to monitor variations in calcium ion homeostasis in vivo after small intestinal I/R injury. The T1 values of the intestinal I/R group were significantly lower (P < 0.05) than that of the control group at 5, 10, and 15 min after Mn(2+) infusion. Our data suggest that MnCl2 has the potential to be an MRI contrast agent that can be effectively used to monitor changes in intracellular Ca(2+) homeostasis during the early stages of intestinal I/R injury.