EMAIL THIS PAGE TO A FRIEND
Advanced materials (Deerfield Beach, Fla.)
A Facile PDMS-Assisted Crystallization for the Crystal-Engineering of C60 Single-Crystal Organic Field-Effect Transistors.
PMID 26088050
Abstract
Poly(dimethylsiloxane) (PDMS)-assisted crystallization (PAC) is a facile method to produce oriented C60 crystal arrays. Changing the drying mechanism from evaporation to solvent absorption (by PDMS) widens the solvent selection and facilitates the engineering of both the macroscopic shape and the microscopic lattice structure of the crystal arrays. The method also shows the potential to be applied to other organic semiconductors and large-area production.