Journal of pharmaceutical sciences
Design and In Vivo Anti-Inflammatory Effect of Ketoprofen Delayed Delivery Systems.
PMID 26088065
For the treatment of inflammatory-based diseases affected by circadian rhythms, the development of once-daily dosage forms is required to target early morning symptoms. In this study, Zn-alginate beads containing ketoprofen (K) were developed by a tandem technique prilling/ionotropic gelation. The effect of main critical variables on particles micromeritics, inner structure as well as on drug loading and in vitro drug release was studied. The inxa0vivo anti-inflammatory efficacy was evaluated using a modified protocol of carrageenan-induced edema in rat paw administering beads to rats by oral gavage at 0, 3, or 5xa0h before edema induction. Good drug loading and desired particle size and morphology were obtained for the optimized formulation F20. In vitro dissolution studies showed that F20 had a gastroresistant behavior and delayed release of the drug in simulated intestinal fluid. The in vitro delayed release pattern was clearly reflected in the prolonged anti-inflammatory effect inxa0vivo of F20, compared to pure ketoprofen; F20, administered 3xa0h before edema induction, showed a significant anti-inflammatory activity, reducing maximum paw volume in response to carrageenan injection, whereas no response was observed for ketoprofen. The designed beads appear a promising platform suitable for a delayed release of anti-inflammatory drugs. © 2015 Wiley Periodicals, Inc. and the American Pharmacists Association J Pharm Sci 104:3451-3458, 2015.