EMAIL THIS PAGE TO A FRIEND
Clinical therapeutics
Susceptibility Profile of Ceftolozane/Tazobactam and Other Parenteral Antimicrobials Against Escherichia coli, Klebsiella pneumoniae, and Pseudomonas aeruginosa From US Hospitals.
PMID 26088525
Abstract
Escherichia coli, Klebsiella pneumoniae, and Pseudomonas aeruginosa are frequently isolated pathogens in the hospital setting, and antimicrobial resistance among these organisms is on the rise. In an attempt to meet the challenge of gram-negative resistance, new therapies, including ceftolozane/tazobactam (C/T), were recently approved by the Food and Drug Administration, and others are in late-stage development. The purpose of this study is to describe the in vitro potency of C/T and other parenteral antimicrobials against a geographically diverse population of E coli, K pneumoniae, and P aeruginosa collected in US hospitals. In 2013 to 2014, 44 hospitals provided nonduplicate, nonurine isolates of E coli (n = 1306), K pneumoniae (n = 1205), and P aeruginosa (n = 1257) from adult inpatients. MICs for C/T and 11 other antimicrobials were determined with broth microdilution methods. The carbapenems, C/T, and colistin displayed the highest percentage of susceptibility and lowest MIC90 against the Enterobacteriaceae, followed by piperacillin/tazobactam (TZP), cefepime, tobramycin, aztreonam, ceftriaxone, and ciprofloxacin. C/T displayed the greatest potency (MIC90 = 2 mg/L) and 97% susceptibility of all compounds against P aeruginosa. In addition, C/T was highly active against P aeruginosa that were nonsusceptible to the carbapenems or TZP or were multidrug resistant and extended-spectrum β-lactamase-producing Enterobacteriaceae. This national survey reported high levels of nonsusceptibility to antimicrobials among both Enterobacteriaceae and P aeruginosa. In contrast, many of these resistant pathogens were susceptible to C/T. These data highlight the enhanced potency of C/T and its potential utility for commonly encountered gram-negative nosocomial pathogens.
Related Materials