EMAIL THIS PAGE TO A FRIEND
Plant science : an international journal of experimental plant biology
Genome-wide analysis of SnRK gene family in Brachypodium distachyon and functional characterization of BdSnRK2.9.
PMID 26089150
Abstract
The sucrose non-fermenting 1 (SNF1)-related protein kinases (SnRKs) play key roles in plant signaling pathways including responses to biotic and abiotic stresses. Although SnRKs have been systematically studied in Arabidopsis and rice, there is no information concerning SnRKs in the new Poaceae model plant Brachypodium distachyon. In the present study, a total of 44 BdSnRKs were identified and classified into three subfamilies, including three members of BdSnRK1, 10 of BdSnRK2 and 31 of BdSnRK3 (CIPK) subfamilies. Phylogenetic reconstruction, chromosome distribution and synteny analyses suggested that BdSnRK family had been established before the dicot-monocot lineage parted, and had experienced rapid expansion during the process of plant evolution since then. Expression analysis of the BdSnRK2 subfamily showed that the majority of them could respond to abiotic stress and related signal molecules treatments. Protein-protein interaction and co-expression analyses of BdSnRK2s network showed that SnRK2s might be involved in biological pathway different from that of dicot model plant Arabidopsis. Expression of BdSnRK2.9 in tobacco resulted in increased tolerance to drought and salt stresses through activation of NtABF2. Taken together, comprehensive analyses of BdSnRKs would provide a basis for understanding of evolution and function of BdSnRK family.