EMAIL THIS PAGE TO A FRIEND
American journal of physiology. Gastrointestinal and liver physiology
Functional characteristics of L1156F-CFTR associated with alcoholic chronic pancreatitis in Japanese.
PMID 26089335
Abstract
Although cystic fibrosis is rare in Japanese, measurement of sweat Cl(-) has suggested mild dysfunction of cystic fibrosis transmembrane conductance regulator (CFTR) in some patients with chronic pancreatitis. In the present study, we have investigated the association of CFTR variants and chronic pancreatitis in Japanese and the functional characteristics of a Japanese- and pancreatitis-specific CFTR variant, L1156F. Seventy patients with alcoholic chronic pancreatitis, 18 patients with idiopathic chronic pancreatitis, and 180 normal subjects participated. All exons and their boundaries and promoter region of the CFTR gene were sequenced. Human embryonic kidney-293 cells were transfected with three CFTR variants (M470V, L1156F, and M470V+L1156F), and the protein expression was examined. Xenopus laevis oocytes were injected with the CFTR variants, and bicarbonate (HCO3 (-)) transport activity was examined. CFPAC-1 cells were transfected with the CFTR variants and Cl(-)/HCO3 (-) exchange activity was examined. Six variants (E217G, I556V, M470V, L1156F, Q1352H, and R1453W) were identified in the coding region of the CFTR gene. Cystic fibrosis-causing mutations were not found. The allele frequencies of L1156F and Q1352H in alcoholic chronic pancreatitis (5.0 and 7.9%) were significantly (P < 0.01) higher than those in normal subjects (0.6 and 1.9%). L1156F was linked with a worldwide CFTR variant, M470V. Combination of M470V and L1156F significantly reduced CFTR expression to ∼60%, impaired CFTR-mediated HCO3 (-)/Cl(-) transport activity to 50-60%, and impaired CFTR-coupled Cl(-)/HCO3 (-) exchange activity to 20-30%. The data suggest that the Japanese-specific CFTR variant L1156F causes mild dysfunction of CFTR and increases the risk of alcoholic chronic pancreatitis in Japanese.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P9417
Palmitoleic acid, ≥98.5% (GC), liquid
C16H30O2
91660
Tridodecylamine, ≥97.0% (GC)
C36H75N