EMAIL THIS PAGE TO A FRIEND
Scientific reports
Novel small-molecule SIRT1 inhibitors induce cell death in adult T-cell leukaemia cells.
PMID 26091232
Abstract
Adult T-cell leukaemia/lymphoma (ATL) is an aggressive T-cell malignancy that develops after long-term infection with human T-cell leukaemia virus (HTLV)-1. The identification of new molecular targets for ATL prevention and treatment is desired. SIRT1, a nicotinamide adenine dinucleotide(+) -dependent histone/protein deacetylase, plays crucial roles in various physiological processes, including aging and apoptosis. We previously reported that ATL patients had significantly higher SIRT1 protein levels than healthy controls. Here, we demonstrate that two novel small-molecule SIRT1 inhibitors, NCO-01/04, reduced cell viability and enhanced apoptotic cells in peripheral blood monocyte cells of patients with acute ATL, which has a poor prognosis. NCO-01/04 also reduced the cell viability with DNA fragmentation, Annexin V-positive cells, and caspase activation. However, a caspase inhibitor did not inhibit this caspase-dependent cell death. NCO-01/04 enhanced the endonuclease G level in the nucleus with loss of the mitochondrial transmembrane potential, which can promote caspase-independent death. Interestingly, NCO-01/04 increased the LC3-II-enriched protein fraction, indicating autophagosome accumulation as well as autophagy. Thus, NCO-01/04 simultaneously caused caspase activation and autophagy. These results suggest that NCO-01/04 is highly effective against ATL cells in caspase-dependent or -independent manners with autophagy, and that its clinical application might improve the prognosis of patients with this fatal disease.
Related Materials