EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences
Chiral analysis of bambuterol, its intermediate and active drug in human plasma by liquid chromatography-tandem mass spectrometry: Application to a pharmacokinetic study.
PMID 26092775
Abstract
A sensitive liquid chromatography-tandem mass spectrometry (LC-MS/MS) method has been developed for simultaneous chiral analysis of an antiasthma drug bambuterol, its key intermediate monocarbamate bambuterol and its active drug terbutaline in human plasma. All samples were extracted with ethyl acetate and separated on an Astec Chirobiotic T column under isocratic elution with a mobile phase consisting of methanol and water with the addition of 20mm ammonium acetate and 0.005% (v/v) formic acid at 0.6mL/min. The analytes were detected by a Xevo TQ-S tandem mass spectrometer with positive electrospray ionization in multiple reaction monitoring mode. The established method has high sensitivity with the lower limit of quantifications of 25.00pg/mL for bambuterol enantiomers, and 50.00pg/mL for monocarbamate bambuterol and terbutaline enantiomers, respectively. The calibration curves for bambuterol enantiomers were linear in the range of 25.00-2500pg/mL, and for monocarbamate bambuterol and terbutaline enantiomers were linear in the range of 50.00-5000pg/mL. The intra- and inter-day precisions were <12.4%. All the analytes were separated in 18.0min. For the first time, the validated method was successfully applied to an enantioselective pharmacokinetic study of rac-bambuterol in 8 healthy volunteers. According to the results, this chiral LC-MS/MS assay provides a suitable and robust method for the enantioselectivity and interaction study of the prodrug bambuterol, the key intermediate monocarbamate bambuterol and its active drug terbutaline in human.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A1542
Ammonium acetate, for molecular biology, ≥98%
C2H7NO2
A7330
Ammonium acetate, BioXtra, ≥98%
C2H7NO2
372331
Ammonium acetate, 99.999% trace metals basis
C2H7NO2
A7262
Ammonium acetate, reagent grade, ≥98%
C2H7NO2
A2706
Ammonium acetate solution, for molecular biology, 7.5 M
C2H7NO2
11018AST Astec® CHIROBIOTIC® V Chiral HPLC Column, 5 μm particle size, L × I.D. 10 cm × 2.1 mm
11019AST Astec® CHIROBIOTIC® V Chiral HPLC Column, 5 μm particle size, L × I.D. 15 cm × 2.1 mm
11020AST Astec® CHIROBIOTIC® V Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 2.1 mm
11010AST Astec® CHIROBIOTIC® V Chiral HPLC Column, 5 μm particle size, L × I.D. 10 cm × 3 mm
11022AST Astec® CHIROBIOTIC® V Chiral HPLC Column, 5 μm particle size, L × I.D. 10 cm × 4.6 mm
11023AST Astec® CHIROBIOTIC® V Chiral HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm
11024AST Astec® CHIROBIOTIC® V Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
11034AST Astec® CHIROBIOTIC® V Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 10 mm
11044AST Astec® CHIROBIOTIC® V Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 21.2 mm
11101AST Astec® CHIROBIOTIC® V Chiral HPLC Guard Column, Column, 5 μm particle size, L × I.D. 2 cm × 1 mm
11100AST Astec® CHIROBIOTIC® V Chiral HPLC Guard Column, Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm
B8684
Bambuterol hydrochloride, >98% (HPLC), powder
C18H30N3O5Cl
270989
Ethyl acetate, anhydrous, 99.8%
C4H8O2
437549
Ethyl acetate, ACS reagent, ≥99.5%
C4H8O2
270520
Ethyl acetate, HPLC grade, 99.8%
C4H8O2
W241407
Ethyl acetate, ≥99%, FCC, FG
C4H8O2
09-0703
Ethyl acetate, for residue analysis, JIS 5000
C4H8O2
09-0730
Ethyl acetate, for HPLC
C4H8O2
W241415
Ethyl acetate, natural, ≥99%, FCC, FG
C4H8O2
537446
Ethyl acetate, ReagentPlus®, ≥99.8%
C4H8O2
L092004
Ethyl acetate
C4H8O2
L092003
Ethyl acetate
C4H8O2
N2126
Neostigmine methyl sulfate
C13H22N2O6S
330361
Soda ash, (99% as sodium carbonate or 58% as sodium oxide), technical grade, light
CNa2O3
S7795
Sodium carbonate, BioXtra, ≥99.0%
CNa2O3