EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical and biomedical analysis
Application of small-angle X-ray scattering to the characterization and quantification of the drug transport nanosystem based on the soybean phosphatidylcholine.
PMID 26093243
Abstract
Phospholipid transport nanosystem (PTNS) for drug delivery is based on soybean phosphatidylcholine. The morphology of PTNS is investigated by means of small-angle X-ray scattering. The obtained results allow one to answer the key question from the viewpoint of organization of drug incorporation whether the PTNS nanoparticles have a structure of micelles or vesicles. It is demonstrated that PTNS is a vesicular system with an average vesicle radius of 160 ± 2Å.