EMAIL THIS PAGE TO A FRIEND
Tissue engineering. Part C, Methods
Effects of Stirring and Fluid Perfusion on the In Vitro Degradation of Calcium Phosphate Cement/PLGA Composites.
PMID 26094637
Abstract
In vitro degradation rates of calcium phosphate bioceramics are investigated using a large variation of soaking protocols that do not all match the dynamic conditions of the perfused physiological environment. Therefore, we studied the effect of stirring and fluid perfusion on the in vitro degradation rate of apatitic calcium phosphate cements (CPC) containing poly(lactic-co-glycolic acid) (PLGA) microspheres. The composites were soaked in phosphate-buffered saline up to 6 weeks under unstirred, stirred, or perfused conditions followed by analysis of mass loss, compression strength, porosity, crystal phase composition, and morphology of the cement composites. The results showed that fluid perfusion reduced the decrease in pH and corresponding degradation rates, while nonperfused soaking conditions (i.e., stirred and unstirred conditions) resulted into more extensive acidification, the rate of which increased with stirring. After 2 weeks, the formation of a secondary brushite phase was observed for cement composites soaked under nonperfused (i.e., stirred and unstirred) conditions, whereas this phase was not detected in cements soaked under perfused conditions. The degradation rate of cement composites decreased in the order unstirred>stirred>perfused, as evidenced by quantification of mass loss, compression strength, and pore morphology. To summarize, we have demonstrated that soaking conditions strongly affected the in vitro degradation process of CPCs. As a consequence, it can be concluded that the experimental design of current in vitro degradation studies does not allow for correlation to (pre-)clinical studies.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
16483
β-tri-Calcium phosphate, puriss. p.a., ≥95% β-phase basis (sintered powder)
Ca3O8P2
C3161
Calcium phosphate tribasic, BioReagent, plant cell culture tested, powder
HCa5O13P3
C5267
Calcium phosphate tribasic, 34.0-40.0% Ca basis
HCa5O13P3
L1250
DL-Lactic acid, 85 % (w/w), syrup
C3H6O3
124737
Glycolic acid, ReagentPlus®, 99%
C2H4O3
G8284
Glycolic acid, BioXtra, ≥98.0% (titration)
C2H4O3
420581
Glycolic acid solution, high purity, 70 wt. % in H2O
C2H4O3
420603
Glycolic acid solution, technical grade, 70 wt. % in H2O
C2H4O3
702153
Hydroxyapatite, nanoparticles, dispersion, 10 wt. % in H2O, <200 nm particle size (BET)
Ca5HO13P3
H0252
Hydroxyapatite, Type I, buffered aqueous suspension
HCa5O13P3
55497
Hydroxyapatite, ′fast flow′
HCa5O13P3
04238
Hydroxyapatite, puriss., meets analytical specification of Ph. Eur., BP, FCC, E341, ≥90% (calculated on glowed substance)
HCa5O13P3
574791
Hydroxyapatite, synthetic, 99.8% trace metals basis (excludes Mg)
HCa5O13P3
21223
Hydroxyapatite, purum p.a., ≥90% (as Ca3(PO4)2, KT)
HCa5O13P3
289396
Hydroxyapatite, reagent grade, powder, synthetic
HCa5O13P3
677418
Hydroxyapatite, nanopowder, <200 nm particle size (BET), ≥97%, synthetic
HCa5O13P3
693863
Hydroxyapatite, nanopowder, <200 nm particle size (BET), contains 5 wt. % silica as dopant, synthetic
HCa5O13P3
W261106
Lactic acid, 85%, FCC
C3H6O3
L6661
Lactic acid, meets USP testing specifications
C3H6O3
W261114
Lactic acid, natural, ≥85%
C3H6O3
252476
Lactic acid solution, ACS reagent, ≥85%
C3H6O3