EMAIL THIS PAGE TO A FRIEND
International journal of clinical and experimental pathology
miR-106a promotes growth and metastasis of non-small cell lung cancer by targeting PTEN.
PMID 26097565
Abstract
MicroRNAs are a class of small non-coding RNAs that play essential roles in cancer development and progression. Recent studies suggested that abnormal expression of miRNAs occurs frequently in non-small cell lung cancer (NSCLC) tissues compared to adjacent normal tissues. In this study, we investigated the expression and the biological roles of miR-106a in non-small cell lung cancer. Our results showed that miR-106a was up-regulated in NSCLC tissues and cell lines. Inhibition of miR-106a in NSCLC cells substantially inhibited cell proliferation, migration, and invasion. Phosphatase and tensin homolog (PTEN) was identified as a direct target of miR-106a, and over-expression of miR-106a suppressed PTEN by direct binding to its 3'-untranslated region (3'-UTR). Furthermore, the presence of miR-106a was inversely correlated with PTEN in NSCLC tissues. Overall, this study suggested that miR-106a inhibited the growth and metastasis of NSCLC cells by decreasing PTEN expression. These data provide novel insights with potential therapeutic applications for the treatment of NSCLC.