EMAIL THIS PAGE TO A FRIEND
Clinical pharmacology in drug development
Pharmacokinetics of Eicosapentaenoic Acid in Plasma and Red Blood Cells After Multiple Oral Dosing With Icosapent Ethyl in Healthy Subjects.
PMID 26097787
Abstract
Icosapent ethyl (IPE) is a prescription form of eicosapentaenoic acid (EPA) ethyl ester. This randomized, open-label study characterized EPA pharmacokinetics. Four healthy subject groups received IPE for 28 days: three received 2 g/day (1 × 1,000 mg BID, 2 × 1,000 mg QD, or 2 × 500 mg BID); one received 4 g/day (2 × 1,000 mg BID) administered with meals. Blood sampling was before the morning dose on days 1, 14, 26, 28, and at specified intervals during an 18-day pharmacokinetic period. EPA was measured in plasma (total and unesterified) and red blood cells (RBCs) by liquid chromatography/tandem mass spectrometry. Mean plasma total EPA increased from 19 µg/mL to a peak (C EPA pharmacokinetic profile demonstrated a slowly cleared, extensively distributed molecule with dose linearity and comparable exposures with BID and QD regimens.