EMAIL THIS PAGE TO A FRIEND
Zhongguo yao li xue bao = Acta pharmacologica Sinica
Biological N-oxidation of piperidine in vitro.
PMID 2609998
Abstract
The biological N-oxidation of piperidine, a pharmacologically active biogenic amine of mammals and human beings, was studied in vitro. After incubation of piperidine-HCl in a fortified rat liver microsomal preparation (9000 x g supernatant) at 37 degrees C for 30 min, 2 metabolites were detected. They were identified as N-hydroxy piperidine and 2, 3, 4, 5-tetrahydro-pyridine-1-oxide as evidenced by TLC, GLC, HPLC, GC-MS and MS.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
56207
1-Hydroxypiperidine, ≥96.0% (GC)
C5H11NO