EMAIL THIS PAGE TO A FRIEND
Advances in experimental medicine and biology
Interconversion between NAD-dependent and O2-dependent types of rat liver xanthine dehydrogenase and difference in kinetic and redox properties between them.
PMID 2610112