EMAIL THIS PAGE TO A FRIEND
Mediators of inflammation
Modulation of Acid Sphingomyelinase in Melanoma Reprogrammes the Tumour Immune Microenvironment.
PMID 26101462
Abstract
The inflammatory microenvironment induces tumours to acquire an aggressive and immunosuppressive behaviour. Since acid sphingomyelinase (A-SMase) downregulation in melanoma was shown to determine a malignant phenotype, we aimed here to elucidate the role of A-SMase in the regulation of tumour immunogenic microenvironment using in vivo melanoma models in which A-SMase was either downregulated or maintained at constitutively high levels. We found high levels of inflammatory factors in low A-SMase expressing tumours, which also displayed an immunosuppressive/protumoural microenvironment: high levels of myeloid-derived suppressor cells (MDSCs) and regulatory T lymphocytes (Tregs), as well as low levels of dendritic cells (DCs). In contrast, the restoration of A-SMase in melanoma cells not only reduced tumour growth and immunosuppression, but also induced a high recruitment at tumour site of effector immune cells with an antitumoural function. Indeed, we observed a poor homing of MDSCs and Tregs and the increased recruitment of CD8(+) and CD4(+) T lymphocytes as well as the infiltration of DCs and CD8(+)/CD44(high) T lymphocytes. This study demonstrates that change of A-SMase expression in cancer cells is sufficient per se to tune in vivo melanoma growth and that A-SMase levels modulate immune cells at tumour site. This may be taken into consideration in the setting of therapeutic strategies.