EMAIL THIS PAGE TO A FRIEND
Dalton transactions (Cambridge, England : 2003)
Simple synthesis of Prussian blue analogues in room temperature ionic liquid solution and their catalytic application in epoxidation of styrene.
PMID 26102295
Abstract
In this article, a new application of the room temperature ionic liquid, tetramethylammonium tetrafluoroborate, is described. This ionic liquid is used as a stabilizing agent and the reaction medium for the synthesis of CoFe Prussian blue analogue nanoparticles using N,N-dimethyl formamide as a complexing agent. The as-synthesized Prussian blue analogues were characterized by various techniques and were used to catalyze the reaction of epoxidation of styrene. The catalytic activity of Prussian blue analogues prepared in ionic liquid was superior to that of Prussian blue analogues prepared in aqueous solution.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
203106
Cobalt(II) nitrate hexahydrate, 99.999% trace metals basis
CoN2O6 · 6H2O