EMAIL THIS PAGE TO A FRIEND
Macromolecular bioscience
Probing the Mucoadhesive Interactions Between Porcine Gastric Mucin and Some Water-Soluble Polymers.
PMID 26102520
Abstract
This study investigates the structural features of porcine gastric mucin (PGM) in aqueous dispersions and its interactions with water-soluble polymers (poly(acrylic acid) (PAA), poly(methacrylic acid) (PMAA), poly(ethylene oxide), and poly(ethylene glycol)) using isothermal titration calorimetry, turbidimetric titration, dynamic light scattering, and transmission electron microscopy. It is established that PAA (450 kDa) and PMAA (100 kDa) exhibit strong specific interactions with PGM causing further aggregation of its particles, while PAA (2 kDa), poly(ethylene oxide) (1 000 kDa), and poly(ethylene glycol) (10 kDa) do not show any detectable effects on mucin. Sonication of mucin dispersions prior to their mixing with PAA (450 kDa) and PMAA (100 kDa) leads to more pronounced intensity of interactions.