Experimental and clinical transplantation : official journal of the Middle East Society for Organ Transplantation
Differentiation of Human-Induced Pluripotent Stem Cells Into Insulin-Producing Clusters by MicroRNA-7.
PMID 26103160
Diabetes results from inadequate insulin production from pancreatic β-cells. Islet cell replacement is an effective approach for diabetes treatment; however, it is not sufficient for all diabetic patients. Thus, finding a new source with effective maturation of β-cells is the major goal of many studies. MicroRNAs are a class of small noncoding ribonucleic acid that regulate gene expression through posttranscriptional mechanisms. MicroRNA-7 has high expression level during pancreatic islet development in humans, thereby playing a critical role in pancreatic β-cell function. We study aimed to develop a protocol to differentiate human-induced pluripotent stem cells efficiently into isletlike cell clusters in vitro by using microRNA-7. Human-induced pluripotent stem cell colonies were transfected with hsa-microRNA-7 by using siPORT NeoFX transfection agent. Total ribonucleic acid was extracted 24 and 48 hours after transfection. The expression of transcription factors which were important during pancreases development was also performed. On the third day, the potency of the clusters was assessed in response to high glucose levels. Diphenylthiocarbazone was used to identify the existence of the β-cells. The presence of insulin and Neurogenin-3 proteins was investigated by immunocytochemistry. Morphologic changes were observed on the first day after chemical transfection, and cell clusters were formed on the third day. The expression of pancreatic specific transcription factors was increased on the first day and significantly increased on the second day. The isletlike cell clusters were positive for insulin and Neurogenin-3 proteins in immunocytochemistry. The clusters were stained with Diphenylthiocarbazone and secreted insulin in a glucose challenge test. MicroRNA-7 transcription factor network is important in pancreatic endocrine differentiation. Chemical transfection with microRNA-7 can differentiate human induced pluripotent stem cells into functional isletlike cell clusters in a short time.
Related Materials
Product #
Molecular Formula
Add to Cart
Zinc isopropylsulfinate, 95%