EMAIL THIS PAGE TO A FRIEND
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine
Cross section measurements of deuteron induced nuclear reactions on natural titanium up to 34 MeV.
PMID 26103622
Abstract
Experimental cross sections for deuteron induced nuclear reactions on natural titanium were measured, using the stacked-foil technique and gamma spectrometry, up to 34 MeV with beams provided by the ARRONAX cyclotron. The experimental cross section values were monitored using the (nat)Ti(d,x)(48)V reaction, recommended by the IAEA. The excitation functions for (nat)Ti(d,x)(44m,46,47,48)Sc are presented and compared with the existing ones and with the TALYS 1.6 code calculations using default models. Our experimental values are in good agreement with data found in the literature. TALYS 1.6 is not able to give a good estimation of the production cross sections investigated in this work. These production cross sections of scandium isotopes fit with the new Coordinated Research Project (CRP) launched by the International Atomic Energy Agency (IAEA) to expand the database of monitor reactions.