EMAIL THIS PAGE TO A FRIEND
Dalton transactions (Cambridge, England : 2003)
Metallosupramolecular Ni2L3 and Ni4L6 complexes of bis-bidentate pyridine-containing ligands: X-ray structures and catalytic proton reduction.
PMID 26104861
Abstract
The pyridine-containing ligands and react with Ni(2+) ions to afford dinuclear triple-stranded helicate and tetrahedral cage supramolecular complexes, respectively; the two architectures exhibit contrasting reactivity towards electro- and photocatalytic proton reduction.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P62003
2-Pyridinecarboxaldehyde, 99%
C6H5NO