EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
Grape Marc Extract-Induced Defense Reactions and Protection against Phytophthora parasitica Are Impaired in NahG Tobacco Plants.
PMID 26105078
Abstract
Grape marc extract (GME) acts as an elicitor of plant defense responses. This study analyzed GME-induced plant defense reactions in NahG transgenic tobacco. Leaf infiltration of NahG leaves revealed HR-like reactions with reduced lesions and weak deployment of autofluorescent compounds in the surrounding infiltrated tissues. The β-1,3-glucanase PR2-, endochitinase PR3-, and osmotin PR5-target transcript levels were strongly lowered in NahG leaves, and the mutant failed to accumulate the antimicrobial PR1 transcripts. GME-induced protection against Phytophthora parasitica var. nicotianae (Ppn) was evaluated on tobacco leaves. The antimicrobial properties of GME against Ppn were evidenced using a range of in vitro tests. GME-sprayed wild-type leaves showed reduced infection areas, whereas GME failed to induce a protective effect against Ppn in NahG leaves. The results suggest that GME-induced plant defense reactions in tobacco plants was mediated by salicylic acid (SA) and that GME-induced protection against Ppn could be the combined result of antimicrobial and defense actions.