EMAIL THIS PAGE TO A FRIEND
Dalton transactions (Cambridge, England : 2003)
Double [3 + 2]-dimerisation cascade synthesis of bis(triazolyl)bisphosphanes, a new scaffold for bidentate bisphosphanes.
PMID 26105114
Abstract
A highly convergent synthesis of bis(triazolylphosphane oxides) was developed by a tandem copper-mediated Huisgen reaction-oxidative coupling. The phosphane oxides were reduced by trichlorosilane and the coordination of the resulting bisphosphanes was studied with various transition metals.