EMAIL THIS PAGE TO A FRIEND
Molecular Oncology
Src family kinases differentially influence glioma growth and motility.
PMID 26105207
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EHU089961 MISSION® esiRNA, esiRNA human LYN (esiRNA1)