EMAIL THIS PAGE TO A FRIEND
Molecular pharmacology
Lysosomal Sequestration Determines Intracellular Imatinib Levels.
PMID 26108972
Abstract
The intracellular uptake and retention (IUR) of imatinib is reported to be controlled by the influx transporter SLC22A1 (organic cation transporter 1). We recently hypothesized that alternative uptake and/or retention mechanisms exist that determine intracellular imatinib levels. Here, we systematically investigate the nature of these mechanisms. Imatinib uptake in cells was quantitatively determined by liquid chromatography-tandem mass spectrometry. Fluorescent microscopy was used to establish subcellular localization of imatinib. Immunoblotting, cell cycle analyses, and apoptosis assays were performed to evaluate functional consequences of imatinib sequestration. Uptake experiments revealed high intracellular imatinib concentrations in HEK293, the leukemic cell lines K562 and SD-1, and a gastrointestinal stromal tumor cell line GIST-T1. We demonstrated that imatinib IUR is time-, dose-, temperature-, and energy-dependent and provide evidence that SLC22A1 and other potential imatinib transporters do not substantially contribute to the IUR of imatinib. Prazosin, amantadine, NH4Cl, and the vacuolar ATPase inhibitor bafilomycin A1 significantly decreased the IUR of imatinib and likely interfere with lysosomal retention and accumulation of imatinib. Costaining experiments with LysoTracker Red confirmed lysosomal sequestration of imatinib. Inhibition of the lysosomal sequestration had no effect on the inhibition of c-Kit signaling and imatinib-mediated cell cycle arrest but significantly increased apoptosis in imatinib-sensitive GIST-T1 cells. We conclude that intracellular imatinib levels are primarily determined by lysosomal sequestration and do not depend on SLC22A1 expression.