EMAIL THIS PAGE TO A FRIEND
FEMS microbiology letters
Cell-to-cell contact and antimicrobial peptides play a combined role in the death of Lachanchea thermotolerans during mixed-culture alcoholic fermentation with Saccharomyces cerevisiae.
PMID 26109361
Abstract
The roles of cell-to-cell contact and antimicrobial peptides in the early death of Lachanchea thermotolerans CBS2803 during anaerobic, mixed-culture fermentations with Saccharomyces cerevisiae S101 were investigated using a commercially available, double-compartment fermentation system separated by cellulose membranes with different pore sizes, i.e. 1000 kDa for mixed- and single-culture fermentations, and 1000 and 3.5-5 kDa for compartmentalized-culture fermentations. SDS-PAGE and gel filtration chromatography were used to determine an antimicrobial peptidic fraction in the fermentations. Our results showed comparable amounts of the antimicrobial peptidic fraction in the inner compartments of the mixed-culture and 1000 kDa compartmentalized-culture fermentations containing L. thermotolerans after 4 days of fermentation, but a lower death rate of L. thermotolerans in the 1000 kDa compartmentalized-culture fermentation than in the mixed-culture fermentation. Furthermore, L. thermotolerans died off even more slowly in the 3.5-5 kDa than in the 1000 kDa compartmentalized-culture fermentation, which coincided with the presence of less of the antimicrobial peptidic fraction in the inner compartment of that fermentation than of the 1000 kDa compartmentalized-culture fermentation. Taken together, these results indicate that the death of L. thermotolerans in mixed cultures with S. cerevisiae is caused by a combination of cell-to-cell contact and antimicrobial peptides.