EMAIL THIS PAGE TO A FRIEND
Caries research
Effectiveness of a Toothpaste with Low Fluoride Content Combined with Trimetaphosphate on Dental Biofilm and Enamel Demineralization in situ.
PMID 26112176
Abstract
The aim of the present study was to evaluate in situ whether a toothpaste with low fluoride associated with sodium trimetaphosphate (TMP) would provide similar effect to that of a 1,100 ppm F toothpaste. This crossover double-blind study consisted of 4 phases (14 days each), during which 10 volunteers wore oral appliances containing 4 enamel bovine blocks. The cariogenic challenge was performed by the application of a 20% sucrose solution (6×/day). The toothpaste treatments (2×/day) were: placebo, 500 ppm F, 500 ppm F plus 1% TMP, and 1,100 ppm F. At the end, enamel mineral loss and biofilm composition were analyzed. The toothpaste with 500 ppm F plus 1% TMP showed the lowest mineral loss (p < 0.05). Regarding the fluoride and calcium concentrations in the enamel and in the biofilm, there were no significant differences between 500 ppm F plus 1% TMP, and 1,100 ppm F toothpastes (p > 0.569), but they were significantly different when compared to toothpaste with 500 ppm F (p < 0.050). The addition of 1% TMP to a low-fluoride toothpaste reduces enamel demineralization in situ similar to a 1,100 ppm F toothpaste.