EMAIL THIS PAGE TO A FRIEND
Molecular human reproduction
Characterization of FSH signalling networks in bovine cumulus cells: a perspective on oocyte competence acquisition.
PMID 26113519
Abstract
Understanding the mechanisms regulating oocyte developmental competence is essential to enhance the clinical efficiency of assisted reproduction. FSH orchestrates the acquisition of oocyte competence, both in vivo and in vitro. Multiple pathways are implicated in FSH signalling; however, their precise coordination remains unresolved. A robust system to investigate FSH signalling is oocyte in vitro maturation (IVM) and we have previously demonstrated better bovine embryo development after FSH addition for the first 6 h during IVM. Using this model, we investigated FSH signalling in cumulus through transcriptomic and pharmacological tools. We demonstrate modulation of cumulus transcriptome by FSH mainly through protein kinase A (PKA) and epidermal growth factor (EGF) pathways. Differentially expressed transcripts were implicated in cumulus expansion, steroidogenesis, cell metabolism and oocyte competence. FSH required rouse-sarcoma oncogene (SRC) for EGF receptor transactivation. PKA and EGF pathway crosstalk was investigated using extracellular signal-regulated kinases (ERK1/2) phosphorylation as the functional end-point. FSH enhanced ERK1/2 activation by the EGF pathway with a simultaneous diminution through PKA. More specifically, FSH increased dual specific phosphatase (DUSP1) transcripts via PKA although DUSP1 protein did not change since EGF was required to prevent degradation. Our findings implicate FSH in PKA and EGF pathway activation, which interact to maintain appropriate levels of ERK1/2 phosphorylation and eventually cumulus expansion, metabolism and steroidogenesis. Moreover, considering the implication of the EGF pathway in GDF9 and BMP15 actions, our findings suggest that FSH may have a role in modulation of the cumulus response to oocyte-secreted factors. This information has implications for improvement of IVM and hence oocyte developmental competence.