EMAIL THIS PAGE TO A FRIEND
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine
An emergency bioassay method for (210)Po in urine.
PMID 26115206
Abstract
A rapid method was developed to efficiently measure (210)Po in urine samples in an emergency situation. Polonium-210 in small urine samples (10 mL) was spontaneously deposited on a stainless steel disc in 1 M HCl at room temperature for 4 h in a polyethylene bottle. The metallic disc was then counted for 4 h by alpha spectrometry. The developed method allowed the preparation of large sample batch in a short time. The method meets the requirements for an emergency bioassay procedure.