EMAIL THIS PAGE TO A FRIEND
Experimental parasitology
Nitric oxide production increases during Toxoplasma gondii encephalitis in mice.
PMID 26115941
Abstract
Toxoplasma gondii is an intracellular parasite with the potential of causing severe encephalitis among immunocompromised human and animals. The aim of this experimental study was to investigate the immunomodulatory and immunopathological role of nitric oxide (NO) in central nervous systems and to identify any correlation between toxoplasmosis neuropathology and investigate the consequences of the cellular responses protect against T. gondii. Mice were infected with ME49 strain T. gondii and levels of endothelial, neuronal and inducible nitric oxide synthase (eNOS, nNOS, iNOS), glial fibrillary acidic protein (GFAP) and neurofilament (NF) were examined in brain tissues by immunohistochemistry, during the development and establishment of a chronic infection at 10 30 and 60 days post infection. Results of the study revealed that the levels of eNOS (pxa0